Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nol-2
Entrez Gene ID
173437
Gene Name
protein-coding
Gene Aliases
CELE_W07E6.1
Chromosome Location
Chr.II: 471240 - 475940 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001316867.3 NP_001303796.1 2 - 3 535 77
NM_061341.6 NP_493742.1 5 - 6 1299 77

View Full Product Details