Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
prp-21
Entrez Gene ID
173440
Gene Name
yeast PRP (splicing factor) related
Gene Aliases
CELE_W07E6.4
Chromosome Location
Chr.II: 484866 - 490403 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061345.10 NP_493746.1 2 - 3 394 67
GenBank mRNA U09415.1 - 2 - 3 405 67

View Full Product Details