Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lgc-34
Entrez Gene ID
173443
Gene Name
Neur_chan_LBD domain-containing protein
Gene Aliases
CELE_T27A1.4
Chromosome Location
Chr.II: 526574 - 530374 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061351.7 NP_493752.1 3 - 4 616 82

View Full Product Details