Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sdz-24
Entrez Gene ID
173457
Gene Name
OB domain-containing protein
Gene Aliases
CELE_K07E8.3
Chromosome Location
Chr.II: 658667 - 659831 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061380.7 NP_493781.1 2 - 3 343 87

View Full Product Details