Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fbxc-28
Entrez Gene ID
187668
Gene Name
F-box C protein
Gene Aliases
CELE_R07C3.6
Chromosome Location
Chr.II: 923821 - 925196 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061451.3 NP_493852.1 1 - 2 126 93

View Full Product Details