Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sdz-4
Entrez Gene ID
183119
Gene Name
SKN-1 Dependent Zygotic transcript
Gene Aliases
CELE_C32B5.16
Chromosome Location
Chr.II: 958918 - 960179 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061464.2 NP_493865.1 1 - 2 111 89

View Full Product Details