Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fbxc-18
Entrez Gene ID
183117
Gene Name
protein-coding
Gene Aliases
CELE_C32B5.14
Chromosome Location
Chr.II: 965575 - 967062 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061467.3 NP_493868.1 1 - 2 177 136

View Full Product Details