Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nas-27
Entrez Gene ID
188809
Gene Name
Zinc metalloproteinase nas-27
Gene Aliases
CELE_T23F4.4
Chromosome Location
Chr.II: 1175925 - 1179323 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061525.4 NP_493926.2 5 - 6 1120 70

View Full Product Details