Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lin-42
Entrez Gene ID
173503
Gene Name
protein-coding
Gene Aliases
CELE_F47F6.1
Chromosome Location
Chr.II: 1233235 - 1252359 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027006.6 NP_001022177.1 2 - 3 390 84
NM_001027007.3 NP_001022178.1 2 - 3 390 84
NM_001322704.3 NP_001309559.1 3 - 4 541 84
NM_001322707.3 NP_001309561.1 3 - 4 541 84
GenBank mRNA AF183400.1 - 2 - 3 390 84

View Full Product Details