Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
jip-1
Entrez Gene ID
173513
Gene Name
protein-coding
Gene Aliases
CELE_F56D12.4
Chromosome Location
Chr.II: 1326989 - 1350136 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001377691.1 NP_001364697.1 5 - 6 797 84
NM_001377692.2 NP_001364548.1 5 - 6 803 84

View Full Product Details