The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001038242.6 | NP_001033331.1 | 5 - 6 | 922 | 61 |
NM_001038243.6 | NP_001033332.1 | 3 - 4 | 460 | 61 | |
NM_001377691.1 | NP_001364697.1 | 13 - 14 | 2365 | 61 | |
NM_001377692.2 | NP_001364548.1 | 13 - 14 | 2356 | 61 | |
NM_001377693.2 | NP_001364549.1 | 5 - 6 | 937 | 61 | |
NM_001377694.2 | NP_001364547.1 | 3 - 4 | 475 | 61 |