Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
jip-1
Entrez Gene ID
173513
Gene Name
protein-coding
Gene Aliases
CELE_F56D12.4
Chromosome Location
Chr.II: 1326989 - 1350136 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038242.6 NP_001033331.1 8 - 9 1320 127
NM_001038243.6 NP_001033332.1 6 - 7 858 127
NM_001377691.1 NP_001364697.1 16 - 17 2763 127
NM_001377692.2 NP_001364548.1 16 - 17 2754 127
NM_001377693.2 NP_001364549.1 8 - 9 1335 127
NM_001377694.2 NP_001364547.1 6 - 7 873 127

View Full Product Details