Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srw-97
Entrez Gene ID
191118
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_ZC204.15
Chromosome Location
Chr.II: 1667522 - 1669013 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061633.1 NP_494034.1 3 - 4 818 87

View Full Product Details