Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- fbxb-19
- Entrez Gene ID
-
186514
- Gene Name
- F-box domain-containing protein
- Gene Aliases
-
CELE_F58E1.9
- Chromosome Location
-
Chr.II: 1688181 - 1689181 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_061641.2 |
NP_494042.1 |
1 - 2 |
93 |
91 |
- Gene Symbol
- fbxc-22
- Entrez Gene ID
-
173538
- Gene Name
- F-box C protein
- Gene Aliases
-
CELE_F58E1.10
- Chromosome Location
-
Chr.II: 1688169 - 1691111 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_061642.6 |
NP_494043.2 |
3 - 4 |
1920 |
91 |
View Full Product Details