Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fbxb-47
Entrez Gene ID
186518
Gene Name
F-box domain-containing protein
Gene Aliases
CELE_F58E1.14
Chromosome Location
Chr.II: 1702382 - 1703463 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061647.4 NP_494048.1 1 - 2 89 131

View Full Product Details