Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- math-3
- Entrez Gene ID
-
173547
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_C08F1.4
- Chromosome Location
-
Chr.II: 1775538 - 1778542 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001383792.1 |
|
6 - 7 |
1167 |
121 |
- Gene Symbol
- F36H5.14
- Entrez Gene ID
-
13183744
- Gene Name
- MATH domain-containing protein
- Gene Aliases
-
CELE_F36H5.14
- Chromosome Location
-
Chr.II: 1775534 - 1778517 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001383791.1 |
|
6 - 7 |
1200 |
121 |
View Full Product Details