Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fbxb-10
Entrez Gene ID
188288
Gene Name
F-box domain-containing protein
Gene Aliases
CELE_T08E11.6
Chromosome Location
Chr.II: 1817271 - 1818400 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061682.2 NP_494083.1 1 - 2 159 96

View Full Product Details