Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fbxa-3
Entrez Gene ID
188289
Gene Name
F-box domain-containing protein
Gene Aliases
CELE_T08E11.7
Chromosome Location
Chr.II: 1818889 - 1820874 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061683.4 NP_494084.2 3 - 4 856 79

View Full Product Details