Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
math-40
Entrez Gene ID
353389
Gene Name
MATH (meprin-associated Traf homology) domain containing
Gene Aliases
CELE_T08E11.3
Chromosome Location
Chr.II: 1830632 - 1831803 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061686.4 NP_494087.2 1 - 2 125 115

View Full Product Details