Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
math-10
Entrez Gene ID
182669
Gene Name
MATH (meprin-associated Traf homology) domain containing
Gene Aliases
CELE_C16C4.15
Chromosome Location
Chr.II: 1871542 - 1873348 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061702.3 NP_494103.1 2 - 3 398 161

View Full Product Details