Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
math-31
Entrez Gene ID
186088
Gene Name
MATH domain-containing protein
Gene Aliases
CELE_F52C6.6
Chromosome Location
Chr.II: 1912450 - 1912841 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061719.2 NP_494120.1 1 - 2 117 65

View Full Product Details