Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bath-7
Entrez Gene ID
186092
Gene Name
BTB domain-containing protein
Gene Aliases
CELE_F52C6.10
Chromosome Location
Chr.II: 1919888 - 1921118 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061723.3 NP_494124.1 1 - 2 132 122

View Full Product Details