Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bath-2
Entrez Gene ID
186093
Gene Name
BTB and MATH domain containing
Gene Aliases
CELE_F52C6.11
Chromosome Location
Chr.II: 1921433 - 1922765 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061724.4 NP_494125.2 4 - 5 810 88

View Full Product Details