Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
math-21
Entrez Gene ID
183363
Gene Name
MATH (meprin-associated Traf homology) domain containing
Gene Aliases
CELE_C40D2.3
Chromosome Location
Chr.II: 1994038 - 1995718 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061744.2 NP_494145.1 1 - 2 114 149

View Full Product Details