Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bath-5
Entrez Gene ID
186650
Gene Name
BTB and MATH domain containing
Gene Aliases
CELE_F59H6.11
Chromosome Location
Chr.II: 2030748 - 2031952 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061757.3 NP_494158.1 1 - 2 124 145

View Full Product Details