Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fbxb-6
Entrez Gene ID
184141
Gene Name
FBA_2 domain-containing protein
Gene Aliases
CELE_F07E5.1
Chromosome Location
Chr.II: 2064797 - 2065870 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001377709.1 NP_001364677.1 1 - 2 95 94

View Full Product Details