Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pqn-42
Entrez Gene ID
173600
Gene Name
protein-coding
Gene Aliases
CELE_F53G2.4
Chromosome Location
Chr.II: 2452970 - 2458981 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061876.8 NP_494277.3 3 - 4 359 65
NM_061877.7 NP_494278.3 3 - 4 364 65

View Full Product Details