Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pqn-42
Entrez Gene ID
173600
Gene Name
protein-coding
Gene Aliases
CELE_F53G2.4
Chromosome Location
Chr.II: 2452970 - 2458981 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129141.3 NP_001122613.1 1 - 2 535 85
NM_061876.8 NP_494277.3 6 - 7 1108 85
NM_061877.7 NP_494278.3 6 - 7 1113 85

View Full Product Details