Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bath-13
Entrez Gene ID
185510
Gene Name
BTB and MATH domain containing
Gene Aliases
CELE_F40B1.1
Chromosome Location
Chr.II: 2520842 - 2522060 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061886.2 NP_494287.1 2 - 3 524 128

View Full Product Details