Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sup-9
Entrez Gene ID
173613
Gene Name
Two pore potassium channel protein sup-9
Gene Aliases
CELE_F34D6.3
Chromosome Location
Chr.II: 2684669 - 2687033 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061932.5 NP_494333.1 6 - 7 829 82
GenBank mRNA AF083652.1 - 6 - 7 773 82
AY357729.1 - 6 - 7 843 82

View Full Product Details