Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clec-3
Entrez Gene ID
183396
Gene Name
C-type LECtin
Gene Aliases
CELE_C41H7.7
Chromosome Location
Chr.II: 3017468 - 3019907 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062025.3 NP_494426.2 2 - 3 452 56

View Full Product Details