Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srg-22
Entrez Gene ID
189539
Gene Name
Serpentine receptor class gamma
Gene Aliases
CELE_Y25C1A.10
Chromosome Location
Chr.II: 3091503 - 3093452 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062042.2 NP_494443.2 2 - 3 443 149

View Full Product Details