Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srh-71
Entrez Gene ID
191868
Gene Name
Serpentine Receptor, class H
Gene Aliases
CELE_T10D4.5
Chromosome Location
Chr.II: 3119332 - 3125409 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001381362.1 NP_001368014.1 9 - 10 3156 85

View Full Product Details