Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sri-33
Entrez Gene ID
191908
Gene Name
Serpentine Receptor, class I
Gene Aliases
CELE_Y27F2A.4
Chromosome Location
Chr.II: 3154558 - 3156849 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001377746.1 NP_001364804.1 2 - 3 275 80

View Full Product Details