Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
wht-4
Entrez Gene ID
173671
Gene Name
ABC transporter domain-containing protein
Gene Aliases
CELE_F02E11.1
Chromosome Location
Chr.II: 3278934 - 3281713 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062094.4 NP_494495.3 7 - 8 1090 65

View Full Product Details