Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
btb-16
Entrez Gene ID
173674
Gene Name
BTB domain-containing protein
Gene Aliases
CELE_F39E9.2
Chromosome Location
Chr.II: 3296883 - 3298330 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062098.6 NP_494499.2 3 - 4 663 68

View Full Product Details