Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sdz-32
Entrez Gene ID
190080
Gene Name
SKN-1 Dependent Zygotic transcript
Gene Aliases
CELE_Y49F6B.5
Chromosome Location
Chr.II: 3474044 - 3474772 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001377763.2 NP_001364706.1 1 - 2 149 81

View Full Product Details