Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyh-1
Entrez Gene ID
190078
Gene Name
CYclin H
Gene Aliases
CELE_Y49F6B.1
Chromosome Location
Chr.II: 3505017 - 3510552 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062163.5 NP_494564.2 5 - 6 481 95

View Full Product Details