Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
apd-3
Entrez Gene ID
173701
Gene Name
protein-coding
Gene Aliases
CELE_W09G10.4
Chromosome Location
Chr.II: 3532527 - 3546438 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062169.6 NP_494570.1 2 - 3 206 72

View Full Product Details