Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srh-60
Entrez Gene ID
191866
Gene Name
Serpentine Receptor, class H
Gene Aliases
CELE_W10G11.9
Chromosome Location
Chr.II: 3583702 - 3586089 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062188.3 NP_494589.1 5 - 6 1407 93

View Full Product Details