Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- srx-97
- Entrez Gene ID
-
184672
- Gene Name
- 7TM_GPCR_Srx domain-containing protein
- Gene Aliases
-
CELE_F19B10.7
- Chromosome Location
-
Chr.II: 3667136 - 3671068 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_062208.3 |
NP_494609.1 |
2 - 3 |
205 |
86 |
- Gene Symbol
- srx-98
- Entrez Gene ID
-
184673
- Gene Name
- 7TM_GPCR_Srx domain-containing protein
- Gene Aliases
-
CELE_F19B10.8
- Chromosome Location
-
Chr.II: 3668585 - 3671056 on Build WBcel235
View Full Product Details