Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sea-1
Entrez Gene ID
173710
Gene Name
T-box transcription factor 18
Gene Aliases
CELE_F19B10.9
Chromosome Location
Chr.II: 3671908 - 3673593 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062210.6 NP_494611.1 3 - 4 903 61

View Full Product Details