Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
oac-4
Entrez Gene ID
182701
Gene Name
Acyl_transf_3 domain-containing protein
Gene Aliases
CELE_C17A2.5
Chromosome Location
Chr.II: 3833250 - 3836015 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322725.1 NP_001309481.1 4 - 5 618 177

View Full Product Details