Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bcmo-2
Entrez Gene ID
173733
Gene Name
Beta-Carotene 15,15'-MonoOxygenase
Gene Aliases
CELE_F53C3.12
Chromosome Location
Chr.II: 3908488 - 3910837 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062293.6 NP_494694.2 3 - 4 1051 77

View Full Product Details