Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ttr-33
Entrez Gene ID
259631
Gene Name
Transthyretin-like protein 33
Gene Aliases
CELE_C37C3.13
Chromosome Location
Chr.V: 7836935 - 7837909 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182392.7 NP_872192.1 1 - 2 161 61

View Full Product Details