Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sre-11
Entrez Gene ID
353444
Gene Name
Serpentine Receptor, class E (epsilon)
Gene Aliases
CELE_C12D5.11
Chromosome Location
Chr.V: 7681820 - 7683957 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182415.5 NP_872215.2 2 - 3 253 96

View Full Product Details