Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
grl-18
Entrez Gene ID
353441
Gene Name
Ground-like domain-containing protein
Gene Aliases
CELE_T05C3.4
Chromosome Location
Chr.V: 5485631 - 5488866 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182425.4 NP_872225.3 7 - 8 761 74

View Full Product Details