Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srg-42
Entrez Gene ID
353468
Gene Name
Serpentine receptor class gamma
Gene Aliases
CELE_T19C4.9
Chromosome Location
Chr.V: 11156941 - 11158047 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001373107.2 NP_001359659.1 3 - 4 288 139

View Full Product Details