Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-16
Entrez Gene ID
173781
Gene Name
Nuclear hormone receptor family member nhr-16
Gene Aliases
CELE_T12C9.6
Chromosome Location
Chr.II: 4438805 - 4440874 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062384.7 NP_494785.1 3 - 4 714 107
GenBank mRNA AF083231.1 - 3 - 4 652 107

View Full Product Details