Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp-23A1
Entrez Gene ID
173787
Gene Name
CYtochrome P450 family
Gene Aliases
CELE_B0304.3
Chromosome Location
Chr.II: 4512211 - 4516136 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062396.6 NP_494797.1 4 - 5 424 75

View Full Product Details